Order Entry
Norway
ContactUsLinkComponent
 
 
 
 
Acitretin ≥98% (by TLC)
  ENZOBMLGR1080010
 :  
undefined
 
 
 
Acitretin ≥98% (by TLC)
  ENZOBMLGR1080010
 :  

 

  • Pk:
    10 mg
  • Seq:
    2275

 

 

  • Purity:
    ≥98% (TLC)

 

 

 :
 :326.4 g/mol
 :Freezer
 :3077
 :9,III
 :GHS07
 :GHS08
 :GHS09
 :55079-83-9
 :C₂₁H₂₆O₃
 :MFCD00866632