Order Entry
Norway
ContactUsLinkComponent
SB202190 ≥98% (by TLC)
SB202190 ≥98% (by TLC)
Catalog # ENZOBMLEI2940001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
SB202190 ≥98% (by TLC)
Catalog # ENZOBMLEI2940001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :331.3 g/mol
Storage Temperature:Freezer
CAS Number:152121-30-7
Formula:C₂₀H₁₄FN₃O
MDL Number:MFCD00941964