L-Buthionine sulphoximine ≥97% (by TLC)
Catalog # ENZOBMLFR1170500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:222.303 g/mol
Storage Temperature:Ambient
CAS Number:83730-53-4
Formula:C₈H₁₈N₂O₃S
MDL Number:MFCD00067000