Order Entry
Norway
ContactUsLinkComponent
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
Catalog # ENZOBMLL1340001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
Catalog # ENZOBMLL1340001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :537.675 g/mol
Storage Temperature:Freezer
CAS Number:84062-61-3
Formula:C₂₆H₅₂NO₈P
MDL Number:MFCD00798242