24(S)-Hydroxycholesterol ≥98% (by TLC), white solid
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:474-73-7
:
Formula:C₂₇H₄₆O₂
:402.663 g/mol
Melting Pt:177…180 °C
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (TLC)