Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9-cis-Retinoic acid
9-cis-Retinoic acid ≥98% (by HPLC)
9-cis-Retinoic acid ≥98% (by HPLC)
Irritant
Health
9-cis-Retinoic acid ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Irritant
Health
9-cis-Retinoic acid ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
5300-03-8
:
Formula:
C₂₀H₂₈O₂
MDL Number:
MFCD00270072
:
300.442 g/mol
:
GHS07
:
GHS08
Specification Test Results
Purity:
≥98% (HPLC)
Specification Test Results