Order Entry
Norway
ContactUsLinkComponent
Raloxifene hydrochloride ≥98% (by TLC)
Raloxifene hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLGR2430050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Raloxifene hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLGR2430050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :510.05 g/mol
Storage Temperature:Freezer
CAS Number:82640-04-8
Formula:C₂₈H₂₇NO₄S
MDL Number:MFCD01938233