Order Entry
Norway
ContactUsLinkComponent
α-Linolenic acid ≥99% (by GC)
α-Linolenic acid ≥99% (by GC)
Catalog # ENZOBMLFA0051000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
α-Linolenic acid ≥99% (by GC)
Catalog # ENZOBMLFA0051000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Purity:
    ≥99% (GC)

Specification Test Results

About this item

 :
 :278.44 g/mol
CAS Number:463-40-1
Formula:C₁₈H₃₀O₂
MDL Number:MFCD00065720