Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
γ-Linolenic acid
γ-Linolenic acid ≥99.0% (by GC), colourless oil
γ-Linolenic acid ≥99.0% (by GC), colourless oil
γ-Linolenic acid ≥99.0% (by GC), colourless oil
ENZOBMLFA0061000
:
ENZO LIFE SCIENCES
:
γ-Linolenic acid ≥99.0% (by GC), colourless oil
ENZOBMLFA0061000
:
ENZO LIFE SCIENCES
:
Pk:
1 g
Seq:
3047
Purity:
≥99% (GC) and one spot by TLC
:
:
278.44 g/mol
:
Freezer
:
506-26-3
:
C₁₈H₃₀O₂
:
MFCD00065718