Order Entry
Norway
ContactUsLinkComponent
Vacu-vials® DR900 Adapter
Vacu-vials® DR900 Adapter
Catalog # CHETA-0215
Leverandør:  CHEMetrics
New Product
Vacu-vials® DR900 Adapter
Catalog # CHETA-0215
Leverandør:  CHEMetrics
New Product
Spesifikasjoner
  • Description:
    Vacu-vials® adapter, 13 mm
  • Pk:
    1
Spesifikasjoner
Om dette produktet
The DR900 Vacu-vials® Adapter is required to use CHEMetrics Vacu-vials® ampoules with the DR900 colorimeter. This adapter is used in conjunction with the 16 mm adapter originally supplied with the DR900. Simply insert the Vacu-vials ampoule into the 16 mm adapter and slide the CHETA-0215 over the ampoule. The CHETA-0215 will center the ampoule and act as a light shield.