Order Entry
Norway
ContactUsLinkComponent
Refractive Index Liquids, Series M
Refractive Index Liquids, Series M
Catalog # CARG1815Y1.750
Supplier:  CARGILLE
CAS Number:  
Refractive Index Liquids, Series M
Catalog # CARG1815Y1.750
Supplier:  CARGILLE
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Refractive index:
    1,750 nD

Specifications

About this item

Refractive index liquids have a broad range of applications in diversified fields.

  • Light yellow liquid
  • Slight unpleasant odour

Adjustment: ±0,0005

Choose from range nD 1,705 to 1,800