Order Entry
Norway
ContactUsLinkComponent
 
Cholesterol (from lanolin (wool fat)), powder Ph. Eur., BP, JP, NF, extra pure, SAFC®
Catalog # 1.03672.5000
CAS Number:  
Cholesterol (from lanolin (wool fat)), powder Ph. Eur., BP, JP, NF, extra pure, SAFC®
Catalog # 1.03672.5000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :386.66 g/mol
Merck Index:13,02221
Boiling Pt:360 °C (1013 hPa)
Melting Pt:147…150 °C
Density:1.07 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:200-353-2
CAS Number:57-88-5
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646