Cholesterol
Katalog # WAKO413-43995
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:280 mL
- Seq:2620
Specifications
About this item
:
:386.66 g/mol
Merck Index:13,02221
Boiling Pt:360 °C (1013 hPa)
Melting Pt:147…150 °C
Density:1.07 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:200-353-2
CAS Number:57-88-5
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646