Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitro-p-toluonitrile
2-Nitro-p-toluonitrile
2-Nitro-p-toluonitrile
2-Nitro-p-toluonitrile
Supplier:
Thermo Scientific
Certificates
Utvid bildet
2-Nitro-p-toluonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
26830-95-5
:
Formula:
C₈H₆N₂O₂
MDL Number:
MFCD00056107
:
162.15
EINECS:
248-019-5
UN:
3439
ADR:
6.1,III