Order Entry
Norway
ContactUsLinkComponent
Accessories for e-Lift
Accessories for e-Lift
Catalog # HEID509-00130-00
Accessories for e-Lift
Catalog # HEID509-00130-00
Specifications
  • For:
    e-Lift lab jack
  • Description:
    e-Lift accessories set, includes a support rod extension (250 mm) for the adaptation of temperature sensors (for example) as well as an adjustable end position for limiting the travel distance
  • Pk:
    1