Order Entry
Norway
ContactUsLinkComponent
Sodium chloride, solid
Sodium chloride, solid
Catalog # CMPLVS1508.80G
CAS Number:  
Sodium chloride, solid
Catalog # CMPLVS1508.80G
CAS Number:  

Specifications

  • Pk:
    80 g
  • Seq:
    3392

Specifications

Specification Test Results

  • pH:
    6 to 9
  • Solubility:
    Water at 20 °C: 358 g/L
  • Assay:
    100,02% ±0,34% (w/w)

Specification Test Results

About this item

 :
 :58.44 g/mol
Boiling Pt:1461 °C
Melting Pt:801 °C
Density:2,16 g/cm³
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477