VITREX® Ultrasafe Safety Lancet
MODU397413
- Application:Diabetes clinics/hospital wards for blood glucose monitoring with increased blood flow
- Colour code:Blue
- Description:Safety lancet, 1,8 mm penetration depth, 21G needle
- Pk:100
Vitrex Ultrasafe lancet is intended for blood glucose and other in vitro diagnostic tests.
- Ergonomically optimised for better handling
- High quality silicone coated needle; less pain and smooth penetration
- Easy targeting of puncture site
- Shelf life of five years
Health workers worldwide face accidental needle-stick injury while caring for patients, putting themselves at risk to bloodborne illnesses including the AIDS virus and Hepatitis.