Anti-TfR (Transferrin R) Mouse Monoclonal Antibody [clone: TFRC/1817]
Catalog # NOVUNBP2-54467
Supplier: Novus Biologicals
Specifications
- Antigen name:TfR (Transferrin R)
- Antigen symbol:TFRC
- Clonality:Monoclonal
- Clone:TFRC/1817
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Format:Azide and BSA free
- Antigen synonyms:TfR (Transferrin R)|TRFR|T9|TFR1|CD71 antigen|transferrin receptor (p90, CD71)|transferrin receptor protein 1|CD71|CD71TRFR|p90|TfR (TransferrinR)|TFRC|TfR|TR
- Purity:Protein A or G purified.
- Storage buffer:10 mM PBS.
- Storage temperature:Store at –20...–80 ℃. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant extracellular fragment (around aa 94-212) of human TfR (Transferrin R) protein (exact sequence is proprietary).
- Pk:100 µG
Specifications
About this item
This antibody is useful in studies of dividing haematopoietic and tumour cell populations, and metabolic activity.
Type:
Antigen: TfR (Transferrin R)
Clonality: Monoclonal
Clone: TFRC/1817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human