Cynomolgus monkey Recombinant Complement Component C5a Protein (from E. coli)
Catalog # PRSI11-356
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pack type:Vial
- Pk:0,1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Cynomolgus monkey
- Storage conditions:Lyophilised protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:102125658
- Protein synonyms:Complement Component 5a|C5a
- Protein/peptide name:C5
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:8,4 kDa
- Sequence:Met 678 - Arg 751
- Formulation:Lyophilized. PBS, pH 7,4.
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Pk Info:0,1 mg