Sodium hydroxide 1/28 mol/l (1/28 N) in aqueous solution
Catalog # LBSE790933
Supplier: LABSERVICE
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548