Boron tribromide 1 M in heptane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-33-4
:
Formula:BBr₃
MDL Number:MFCD00011312
:250.52 g/mol
Boiling Pt:90 °C
Density:0.869 g/cm³
Flash Pt:–1 °C
Storage Temperature:Ambient
EINECS:233-657-9
UN:3384
ADR:6.1,
:GHS06
:GHS08
:GHS05
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear yellow to amber Solution
- Strength:0.9 to 1.1 M