Boron tribromide 1 M in methylene chloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-33-4
:
Formula:BBr₃
MDL Number:MFCD00011312
:250.52 g/mol
Storage Temperature:Ambient
EINECS:233-657-9
UN:3390
ADR:6.1,
:GHS06
:GHS08
:GHS05
Specification Test Results
- Appearance:Clear yellow to dark brown Solution (May darken on storage. May develop some turbidity or precipitate)
- Strength:0.95 to 1.10 M