Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nonynoic acid methyl ester
2-Nonynoic acid methyl ester ≥98%
2-Nonynoic acid methyl ester ≥98%
B21409.14
:
Thermo Fisher Scientific
:
2-Nonynoic acid methyl ester ≥98%
B21409.14
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
25 g
:
111-80-8
:
:
C₁₀H₁₆O₂
:
168.24 g/mol
:
121 °C (20 mmHg)
:
0.915
:
104 °C (219 °F)
:
203-909-2
:
GHS07
:
Ambient
:
MFCD00009547