Iron(III) citrate hydrate Fe(III) 16,5-20% (Fe(II) ≤ 5%)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:334024-15-6
:207399-12-0
Formula:C₆H₅FeO₇·xH₂O
MDL Number:MFCD00149615
:244.948 g/mol (anhy)
Storage Temperature:Romtemperatur
EINECS:222-536-6