2-Chloronicotinic acid
Catalog # EHERC11451900
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:157.56 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:181 °C
EINECS:220-937-0
:GHS07
Storage Temperature:Ambient
CAS Number:2942-59-8
Formula:C₆H₄ClNO₂
MDL Number:MFCD00006236
Boiling Pt:284 °C (1013 hPa)