4,4'-Isopropylidenediphenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80-05-7
:
Formula:C₁₅H₁₆O₂
MDL Number:MFCD00002366
:228.29 g/mol
Merck Index:13,01296
Boiling Pt:220 °C (4 mmHg)
Melting Pt:155…158 °C
Density:1,205 g/cm³ (20 °C)
Flash Pt:227 °C
EINECS:201-245-8
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Flakes or prills
- Infrared spectrum:Conforms
- Melting point:155 to 160 °C
- GC:≥96.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Acidity/alkalinity:≤0.00025 meq/g
- Aluminium (Al):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Chromium (Cr):≤0.05 ppm
- Cobalt (Co):≤0.1 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.2 ppm
- Nickel (Ni):≤0.05 ppm
- Heavy metals (as Pb):≤5 ppm
- Sulfur compounds (as S):≤0.003 %