4-Amino-m-cresol
Catalog # EHERC10204955
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:123.15 g/mol
Boiling Pt:259 °C (1013 hPa)
Melting Pt:177…179 °C
EINECS:220-621-2
CAS Number:2835-99-6
Formula:C₇H₉NO
MDL Number:MFCD00007871