Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lights
Halogen Lights, Lamps and Bulbs
Halogen lamp 12 V
Halogen lamp 12 V
Halogen lamp 12 V
Catalog # 632-0197
Supplier:
OSRAM
Expand image
Halogen lamp 12 V
Catalog # 632-0197
Supplier:
OSRAM
Specifications
Description:
Spare halogen bulb
Base:
G4
Pk:
1
Power:
20 W
Voltage:
12 V
Specifications
About this item
Product Details
Product Details