Order Entry
Norway
ContactUsLinkComponent
Potassium iodate-iodide concentrate for dilution to 0,05 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Potassium iodate-iodide concentrate for dilution to 0,05 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4902.
Supplier:  Avantor
CAS Number:  
Potassium iodate-iodide concentrate for dilution to 0,05 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4902.
Supplier:  Avantor
CAS Number:  

Specification Test Results

  • Concentration after dilution to 1000 ml in 20°C:
    c(I2)=0,05mol/l +/- 0,2 %

Specification Test Results

About this item