Tripod plate, H-foot, PASS
765-1140
: PHYWE
- Description:H-foot, PASS
- Pk:1
Extremely stable H-foot with 5 attachment points and many combination options. Intended for tripod rods 4 to 12 mm or square tripod rods with a cross section of 4×4 mm to 12×12 mm. If a tripod rod is mounted between two standing feet, a compact, stable and easily movable physics bench is obtained.
Dimensions: 193×178×36 mm