Lens with variable focal length
Catalog # 763-0325
Supplier: IMBE HANDELSBOGALET
Specifications
- Description:Lens with variable focal length
- Pk:1
Specifications
About this item
Lens made of silicone rubber, mounted in a screen. The shape can be varied steplessly from convex to concave by changing the water volume in the lens. If an object is placed in front of the lens, the object can be projected on a posterior screen (retina). The focus is adjusted with the volume of water in the lens.