Wire beam tube e / m
Catalog # 765-1206
Supplier: PHYWE
Specifications
- Description:Wire beam tube e / m
- Pk:1
Specifications
About this item
Ice cream sphere filled with noble gas of low pressure. The tube's two cylindrical necks are protected by plastic sleeves and are designed to fit in Helmholtz coils 765-1207. The tube is used to show how an electron beam is deflected in a magnetic field. If the tube is in a homogeneous magnetic field with sufficient strength, the electron beam becomes circular. The tube is equipped with an internal measuring scale with fluorescent markings at distances of 4, 6, 8 and 10 cm (2r) from the cathode. The tube is well suited for determining the charge-mass ratio of the electron with great accuracy.
- Anode voltage: 1,500-5,000 V DC
Suitable voltage source 765-0820.