Order Entry
Norway
Orders LinkContactUsLinkComponent
Coil spring
Coil spring
Marketsource
Supplier:   PHYWE
Coil spring
Supplier:  PHYWE
Marketsource
Certificates
Coil spring
3 Options Available Below
Product Details & Documents
Coil springs made of spring steel. With stirrups at both ends. Suitable for Hook's team and swing attempts.
Product Family Options
Product Information
  • Availability
    Price
  • 765-1151
    Spiral spring 3 N/m
    Spiral spring 3 N/m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 255,00
    kr 255,00
    1 * 1 Items
       kr 255,00
    Specifications:
    Marketsource
    Marketsource
  • 765-1152
    Spiral spring 20 N/m
    Spiral spring 20 N/m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 165,00
    kr 165,00
    1 * 1 Items
       kr 165,00
    Specifications:
    Marketsource
    Marketsource
  • 765-1153
    Spiral spring 30 N/m
    Spiral spring 30 N/m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    Marketsource
    Marketsource