Order Entry
Norway
Orders LinkContactUsLinkComponent
Pendulum bobs
Pendulum bobs
Marketsource
Supplier:   PHYWE
Pendulum bobs
Supplier:  PHYWE
Marketsource
Certificates
Pendulum bobs
3 Options Available Below
Product Details & Documents
Suitable for use as pendulum bobs for frequency experiments. Hardened and polished ball bearings with eyelet.
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • 765-1154
    Steel pendulum bob
    Steel pendulum bob
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    Marketsource
    Marketsource
  • 765-1155
    Steel pendulum bob
    Steel pendulum bob
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 390,00
    kr 390,00
    1 * 1 Items
       kr 390,00
    Specifications:
    Marketsource
    Marketsource
  • 765-1156
    Steel pendulum bob
    Steel pendulum bob
    32 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 790,00
    kr 790,00
    1 * 1 Items
       kr 790,00
    Specifications:
    Marketsource
    Marketsource