Leclanche element
Leclanche element
Catalog # 765-0200
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Used in electrochemistry. Voltage, max 1.5 V. The unit is divisible and consists of glass beaker, cylindrical zinc electrode, plastic beaker, carbon electrode, manganese oxide, crocodile clamps and lid. Fill with ammonium chloride solution (20%).
- A suitable voltage indicator is a slightly rotating motor with a propeller
Specifications
- Description:Leclanche element