Order Entry
Norway
ContactUsLinkComponent
Conductor ball
  765-0920
undefined
Conductor ball
  765-0920
  • Description:
    Conductor ball, metallized
  • Pk:
    1
Very light ball of porous plastic. With thin nylon thread for hanging. Used for influence tests, for transferring charge, eg between capacitor plates etc.
  • Diam: 38 mm
  • Weight: 2 g