Experiment kit, electric field lines
Supplier: PHYWE
Certificates
About this item
For demonstration of electric fields between electrodes of different shapes. The electrodes have been applied in thin conductive layers on plexiglass plates and the field line images appear in groats placed in a cuvette with 1 to 2 mm castor oil (the cuvette is placed on the plexiglass plate). The cuvette has an annular electrode in the outer jacket and a 4 mm connection sleeve. The cleaning is easy when electrodes resp. plexiglass plate never comes into contact with the oil. Suitable voltage source: High voltage generator 0 to 25 kV or Van de Graff generator.
- Dimensions: plates 180 × 180 mm, diam cuvette 120 mm