Measurement inserts for resistance measurement
Leverandør: PHYWE
Sertifikat
Om dette produktet
Measurement insert for resistance measurement. Used together with demonstration instruments 765-1247. Clear, fine-grained scale with number markings. Measuring range window where the information automatically follows the measuring range switch. On the back there are measuring range switches, zero adjustment and contact sleeves for printers.