Order Entry
Norway
Orders LinkContactUsLinkComponent
Microvolt amplifier for demonstration instruments
Microvolt amplifier for demonstration instruments
Microvolt amplifier
Catalog # 765-1293
Marketsource
Supplier:   PHYWE
Microvolt amplifier for demonstration instruments
Catalog # 765-1293
Supplier:  PHYWE
Marketsource
Microvolt amplifier for demonstration instruments
Microvolt amplifier for demonstration instruments
Microvolt amplifier
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
For measuring low currents AC/DC and DC voltages AC/DC eg Hall voltages and thermal voltages. High sensitivity and very good electrical stability. Well shielded from disturbing voltages. Overload protected up to 220 V AC/DC. The front is equipped with two scales 0 to 3 and 0 to 10 both with fine division, measuring area window and automatic function indication. The back is equipped with a function switch, measuring range switch, pushbutton for zero setting and steering wheel for electrical zero adjustment. There is also a printer connection.
Measuring ranges: 0 to 30 µV
0 to 100 µV
0 to 1 mV
0 to 10 mV
Specifications
  • Description:
    Microvolt amplifier