Double sleeve, PASS
Catalog # 765-1145
Supplier: PHYWE
Specifications
- Pk:1
Specifications
About this item
Intended for interconnection of cylindrical rods 4 to 18 mm and/or square rods with a cross section of 4×4 to 12×12 mm. The sleeve can also be used for clamping plates with ∼2 to 14 mm thickness.
Dimensions incl screw: 105 × 75 × 35 mm