Order Entry
Norway
Orders LinkContactUsLinkComponent
9-Hydroxyfluorene

9-Hydroxyfluorene

Glass bottle for solids, 25 g
Catalog # MOLEM63880451
Marketsource
Supplier:   Molekula
9-Hydroxyfluorene
Catalog # MOLEM63880451
Supplier:  Molekula
Marketsource
CAS Number:  1689-64-1
9-Hydroxyfluorene

Glass bottle for solids, 25 g

CAS Number:  1689-64-1
Price
kr 1 100,00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  9-Fluorenol, Fluoren-9-ol
  • CAS Number:1689-64-1
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00001135
  • Molecular Weight:182.22 g/mol
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:152…157 °C
  • Density:1,151 g/cm³ (20 °C)
  • EINECS:216-879-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.