Order Entry
Norway
ContactUsLinkComponent
5-Fluorouracil
Toxic
Health
5-Fluorouracil
Catalog # MOLE22050130-25G
Supplier:  Molekula
CAS Number:  
Toxic
Health
5-Fluorouracil
Catalog # MOLE22050130-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :130.08 g/mol
Merck Index:13,04208
Boiling Pt:347 °C (1013 hPa)
Melting Pt:282…283 °C
Storage Temperature:Ambient
EINECS:200-085-6
UN:2811
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:51-21-8
Formula:C₄H₃FN₂O₂
MDL Number:MFCD00006018