Order Entry
Norway
ContactUsLinkComponent
5-Fluorouracil for synthesis, Sigma-Aldrich®
5-Fluorouracil for synthesis, Sigma-Aldrich®
Catalog # 8.18505.0005
Supplier:  Merck
CAS Number:  
5-Fluorouracil for synthesis, Sigma-Aldrich®
Catalog # 8.18505.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    12 g/l (20 °C)
  • Melting Point:
    282 - 283 °C (decomposition)
  • Molar Mass:
    130.08 g/mol
  • Boiling Point:
    190 - 200 °C (0.13 hPa)
  • pH:
    4.3 - 5.3 (10 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :130.08 g/mol
Merck Index:13,04208
Boiling Pt:347 °C (1013 hPa)
Melting Pt:282…283 °C
Storage Temperature:Ambient
EINECS:200-085-6
UN:2811
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
CAS Number:51-21-8
Formula:C₄H₃FN₂O₂
MDL Number:MFCD00006018