Order Entry
Norway
ContactUsLinkComponent
N,N,N-Trimethylanilinium chloride, Sigma-Aldrich®
Toxic
N,N,N-Trimethylanilinium chloride, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
N,N,N-Trimethylanilinium chloride, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    333 g/l (20 °C)
  • Melting Point:
    246 - 247 °C
  • Molar Mass:
    171.66 g/mol
  • Bulk Density:
    540 kg/m3
  • Explosion Limit:
    1 - 6 %(V)
  • pH:
    4.5 (333 g/l, H2O, 20 °C)

Specification Test Results

Related Information