Order Entry
Norway
ContactUsLinkComponent
 
 
 
 
1,4-Dimethylpiperazine for synthesis, Sigma-Aldrich®
  8.03580.0100
 :  Merck
 :  
 
 
 
1,4-Dimethylpiperazine for synthesis, Sigma-Aldrich®
  8.03580.0100
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Solubility:
    (20 °C) miscible
  • Melting Point:
    -1 °C
  • Molar Mass:
    114.19 g/mol
  • Boiling Point:
    130 - 133 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Flash Point:
    21.5 °C
  • Refractive Index:
    1.4463 (20 °C, 589 nm)
  • Explosion Limit:
    1.8 - 9.6 %(V)
  • Density:
    0.85 g/cm3 (20 °C)
  • pH:
    10 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    180 °C

 

 

 :
 :114.19 g/mol
 :131 °C (1013 hPa)
 :–1 °C
 :0.85 g/cm³ (20 °C)
 :Ambient
 :203-412-0
 :2733
 :3 (8),II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS05
 :GHS07
 :106-58-1
 :C₆H₁₄N₂
 :MFCD00006156