α-Terpinene
Catalog # EHERC17322320
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:136.24 g/mol
Boiling Pt:173 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.834 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:202-795-1
UN:2319
ADR:3,III
CAS Number:99-86-5
Formula:C₁₀H₁₆
MDL Number:MFCD00001534