D-(-)-Ribose
Catalog # EHERC16813700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:150.13 g/mol
Merck Index:12,08371
Boiling Pt:331 °C (1013 hPa)
Melting Pt:90…95 °C
Density:0.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-059-4
CAS Number:50-69-1
Formula:C₅H₁₀O₅
MDL Number:MFCD00135453