Mitotane
Catalog # EHERC12030000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:320.04 g/mol
Merck Index:12,06302
Melting Pt:77…78 °C
Storage Temperature:Ambient
EINECS:200-166-6
UN:2761
ADR:6.1,III
:GHS08
CAS Number:53-19-0
Formula:C₁₄H₁₀Cl₄
MDL Number:MFCD00000850