Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Acetyl-5-bromothiophene
2-Acetyl-5-bromothiophene
2-Acetyl-5-bromothiophene
2-Acetyl-5-bromothiophene
Supplier:
Thermo Scientific
Certificates
Expand image
2-Acetyl-5-bromothiophene
Supplier:
Thermo Scientific
About this item
CAS Number:
5370-25-2
:
Formula:
C₆H₅BrOS
MDL Number:
MFCD00014528
:
205.08 g/mol
Boiling Pt:
253 °C (1013 hPa)
Melting Pt:
92…93 °C
EINECS:
226-363-7