Order Entry
Norway
ContactUsLinkComponent
3-Chloro-1H-indazole
3-Chloro-1H-indazole
Catalog # MAYBJFD02345.1
Supplier:  Thermo Scientific
CAS Number:  
3-Chloro-1H-indazole
Catalog # MAYBJFD02345.1
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.58 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:147…148 °C
EINECS:249-444-9
CAS Number:29110-74-5
Formula:C₇H₅ClN₂
MDL Number:MFCD00067527